
2,3-Dichloroquinoxaline ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:2213-63-0
:
Formula:C₈H₄Cl₂N₂
MDL Number:MFCD00006720
:199.04 g/mol
Boiling Pt:301 °C (1013 hPa)
Melting Pt:152…153 °C
EINECS:218-667-3
UN:2811
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:White to Light yellow to Dark green powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:151.0 to 154.0 deg-C
- Solubility in Toluene:very faint turbidity
Specification Test Results
Frequently Bought Together