6-Nitropiperonal ≥97.0% (by GC)
Catalog # TCIAN0537-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Light orange to Yellow to Green powder to crystal
- Purity (GC):min. 97.0 %
- Melting point:94.0 to 99.0 deg-C
Specification Test Results
About this item
:
:195.13 g/mol
Melting Pt:98 °C
EINECS:211-926-1
CAS Number:712-97-0
Formula:C₈H₅NO₅
MDL Number:MFCD00005829