

5,6-Epoxy-1-hexene ≥96.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:10353-53-4
:
Formula:C₆H₁₀O
MDL Number:MFCD00010051
:98.14 g/mol
Boiling Pt:121°C
Melting Pt:–156 °C
Density:0,869 g/cm³ (20 °C)
Flash Pt:15°C
Storage Temperature:Refrigerator
EINECS:233-771-9
UN:1993
ADR:3,II
:GHS02
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 96.0 %
Specification Test Results
Frequently Bought Together