

2-Bromobenzoyl chloride ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:7154-66-7
:
Formula:C₇H₄BrClO
MDL Number:MFCD00000655
:219.46 g/mol
Boiling Pt:245°C
Melting Pt:11 °C
Density:1,67 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:230-507-4
UN:3265
ADR:8,III
:GHS05
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together