Order Entry
Italy
ContactUsLinkComponent
6-Bromopiperonal ≥98.0% (by GC)
Irritant
6-Bromopiperonal ≥98.0% (by GC)
Catalog # TCIAB2332-5G
Supplier:  TCI
CAS Number:  
undefined
Irritant
6-Bromopiperonal ≥98.0% (by GC)
Catalog # TCIAB2332-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    128.0 to 132.0 deg-C
  • Solubility in hot Methanol:
    almost transparency

Specification Test Results

About this item

 :
 :229.03 g/mol
 :GHS07
CAS Number:15930-53-7
Formula:C₈H₅BrO₃
MDL Number:MFCD00022952