
4,5-Dibromo-o-xylene ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:24932-48-7
:
Formula:C₈H₈Br₂
MDL Number:MFCD00082743
:263.96 g/mol
Boiling Pt:278°C
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:85.0 to 89.0 deg-C
Specification Test Results
Frequently Bought Together