Cold resistant gloves, ActivArmr® Winter Monkey Grip® 23-173 / 23-191 / 23-193
Supplier: Ansell
Certificates
About this item
These cotton jersey gloves with full PVC coating keep hands warm, dry and protected from abrasions.
- PVC coating provides oil and grease repellency
- PVC coating remains flexible at low temperatures
- Excellent grip
- Antistatic according to EN 1149
Ideal when handling objects coated in grease and oils and when handling chemicals in cold environments. Also suitable for winter transportation of oil and for the maintenance of cool rooms and storage.
Certifications: 23-173: EN 511 (011), EN 388:2016 (3241B), Cat. II
23-191/23-193: EN 511 (111), EN 388:2016 (3121B), Cat. II
23-191/23-193: EN 511 (111), EN 388:2016 (3121B), Cat. II
Specifications
- Catalog No:
- ANSE23-191/10
- ANSE23-173/11
- ANSE23-191/11
- ANSE23-193/11
- ANSE23-193/10
- ANSE23-173/10
- Model:
- 23-191
- 23-173
- 23-191
- 23-193
- 23-193
- 23-173
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- Cotton
- Cotton
- Cotton
- Cotton
- Cotton
- Cotton
- Finish:
- Fully coated
- Fully coated
- Fully coated
- Fully coated
- Fully coated
- Fully coated
- Cuff style:
- Knit wrist
- Safety cuff
- Knit wrist
- Safety cuff
- Safety cuff
- Safety cuff
- EN norm:
- EN 511 (111), EN 388:2016 (3121B)
- EN 511 (011), EN 388:2016 (3241B)
- EN 511 (111), EN 388:2016 (3121B)
- EN 511 (111), EN 388:2016 (3121B)
- EN 511 (111), EN 388:2016 (3121B)
- EN 511 (011), EN 388:2016 (3241B)
- Anti-static:
- +
- +
- +
- +
- +
- +