Order Entry
Italy
ContactUsLinkComponent
Filter plates, 96-well, PreDictor™
Filter plates, 96-well, PreDictor™
Catalog # 28-9258-19
Supplier:  Cytiva
Filter plates, 96-well, PreDictor™
Catalog # 28-9258-19
Supplier:  Cytiva
Specifications
  • No. of wells:
    96
  • Description:
    PreDictor™ Capto adhere, 50 µl
  • Material:
    PP/PE
  • Pk:
    4
Specifications
About this item
PreDictor™ plates are disposable, 96-well filter plates for parallel screening of media and chromatographic conditions such as binding, wash, and elution conditions. Data generated using PreDictor™ plates show good correlation with data from chromatography columns, making the plates an excellent tool for initial screening of process conditions.
  • Supports high throughput process development (HTPD) by allowing parallel screening of chromatographic conditions using a 96-well plate format
  • Allows fast and efficient evaluation of parameters for binding/wash/elution conditions, and media benchmarking
  • Results show good correlation with data obtained by column chromatography
  • Each well is prefilled with defined amounts of chromatography media
  • Can be used with centrifugation or vacuum, manually or in automated robotic systems
  • Assist software supports the PreDictor™ workflow from set-up of experimental design to data evaluation