High vacuum bellow valves, VE series
High vacuum, helium leak-tested right angle valves VE have stainless steel bellows which seals the drive mechanism without any gap on the vacuum side.
- High conductance due to large stroke length
- Excellent gas tightness, minimum degassing
- Stainless steel version VE WIG bakeable to 150 °C
- Simple dismantling and easy changing of seal
- Dimensions of the VE valve are the same as elbow with corresponding nominal width
Frequently Bought Together