2,3-Diphenylpyrazine ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1588-89-2
:
Formula:C₁₆H₁₂N₂
MDL Number:MFCD00053000
Melting Pt:119…122 °C
:232.29 g/mol
Storage Temperature:Ambient
Related Information
Frequently Bought Together