Order Entry
Italy
ContactUsLinkComponent
cis-Dicyclohexano-18-crown-6 (mixture of syn-cis and anti-cis isomers) 98%
Toxic
cis-Dicyclohexano-18-crown-6 (mixture of syn-cis and anti-cis isomers) 98%
Certificates
undefined
Toxic
cis-Dicyclohexano-18-crown-6 (mixture of syn-cis and anti-cis isomers) 98%

Specification Test Results

  • Appearance:
    White to light beige Crystalline solid
  • Infrared spectrum:
    Conforms
  • Melting point:
    47 to 53 °C
  • GC:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together