(S)-(-)-Menthyltoluene-4-sulphinate ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:1517-82-4
:
Formula:C₁₇H₂₆O₂S
MDL Number:MFCD00010192
:294.4521 g/mol
Storage Temperature:Freezer
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:100.0 to 106.0 deg-C
- Specific rotation [a]20/D:-195.0 to -205.0 deg (C=2, Acetone)
Specification Test Results
Frequently Bought Together