Order Entry
Italy
ContactUsLinkComponent
4'-Nitrobenzo-15-crown-5 (4'-nitrobenzo-1,4,7,10,13-pentaoxacyclopentadecane) ≥99%
Irritant
4'-Nitrobenzo-15-crown-5 (4'-nitrobenzo-1,4,7,10,13-pentaoxacyclopentadecane) ≥99%
Catalog # A16225.06
CAS Number:  
Irritant
4'-Nitrobenzo-15-crown-5 (4'-nitrobenzo-1,4,7,10,13-pentaoxacyclopentadecane) ≥99%
Catalog # A16225.06
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

About this item

 :
 :313.30 g/mol
Melting Pt:93…95 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:60835-69-0
Formula:C₁₄H₁₉NO₇
MDL Number:MFCD00060713