Order Entry
Italy
ContactUsLinkComponent
Allyl mercaptan 70% tech. remainder mainly sulfide
Irritant
Flammable
Allyl mercaptan 70% tech. remainder mainly sulfide
Catalog # ACRO153710250
CAS Number:  
undefined
Irritant
Flammable
Allyl mercaptan 70% tech. remainder mainly sulfide
Catalog # ACRO153710250
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • GC:
    ≥65 %

Specification Test Results

About this item

 :
 :74.15 g/mol
Density:0.93 g/cm³
Flash Pt:18 °C
Storage Temperature:Refrigerator
EINECS:212-792-7
UN:1228
ADR:3,II
 :GHS07
 :GHS02
CAS Number:870-23-5
Formula:C₃H₆S
MDL Number:MFCD00004894