2,4,6-Tribromoresorcinol ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2437-49-2
:
Formula:C₆H₃Br₃O₂
MDL Number:MFCD00009717
:346.82 g/mol
Melting Pt:114…116 °C
EINECS:219-442-2
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together