Order Entry
Italy
ContactUsLinkComponent
Organic reference standard, Tetrachlorobisphenol A 50 µg/ml in toluene, CIL
Organic reference standard, Tetrachlorobisphenol A 50 µg/ml in toluene, CIL
Catalog # PROOCILULM76061.2
CAS Number:  
Organic reference standard, Tetrachlorobisphenol A 50 µg/ml in toluene, CIL
Catalog # PROOCILULM76061.2
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1,2 mL
  • CAS No.:
    79-95-8
  • Description:
    Tetrachlorobisphenol A 50 µg/ml in toluene
  • Formula:
    C₁₅H₁₂Cl₄O₂
  • Description:
    Tetrachlorobisphenol A

Specifications

About this item