Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Diethylbenzene (mixture of isomers)
Diethylbenzene (mixture of isomers)
Diethylbenzene (mixture of isomers)
:
Thermo Fisher Scientific
Diethylbenzene (mixture of isomers)
:
Thermo Fisher Scientific
:
25340-17-4
:
:
C₁₀H₁₄
:
MFCD00792900
:
134.22 g/mol
:
182…184 °C
:
0.87
:
55 °C (131 °F)
:
Ambient
:
246-874-9
:
2049
:
3,III
:
GHS02
msds_ALFAA19548
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years
Frequently Bought Together
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
N,N-Dimethylformamide, anhydrous (max. 0.005% H₂O) ≥99.8%
: VWR Chemicals
Petroleum spirit, 40…60 °C, GPR RECTAPUR®
: VWR Chemicals
Glycerine 87% 86%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
1-Propanol, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
: Merck
Petroleum spirit, 40…65 °C, anhydrous, TECHNICAL (max. 0.01% aromatic hydrocarbons)
: VWR Chemicals
Formaldehyde ≥37% in aqueous solution stabilised, GR ACS, Reag. Ph. Eur. analytical reagent, Sigma-Aldrich®
: Merck
Diethyl ether ≥99% stabilised, GPR RECTAPUR®
: VWR Chemicals
Trifluoroacetic acid, Uvasol® for spectroscopy, Supelco®
: Merck
Tetrachloroethylene, Uvasol® for spectroscopy, Supelco®
: Merck
Glycerine (glycerol) USP, Ph. Eur., BP
: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
: VWR Chemicals
Acetonitrile : water : trifluoroacetic acid (50 : 47,5 : 2,5), HiPerSolv CHROMANORM®
: VWR Chemicals
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
: VWR Chemicals