



Diethylbenzene (mixture of isomers) mixture of o-, m- and p-isomers 95% (sum of isomers)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:25340-17-4
:
Formula:C₁₀H₁₄
MDL Number:MFCD00792900
:134.22 g/mol
Boiling Pt:180…182 °C (760 mmHg)
Melting Pt:–73 °C
Density:0.869 g/cm³
Flash Pt:56 °C
EINECS:246-874-9
UN:2049
ADR:3,III
:GHS02
:GHS07
:GHS08
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4940 to 1.4960 (20°C, 589 nm)
- GC:≥95.0 % (sum of o-, m-, p-isomers)
- Titration with HCl:0.37 to 0.43 M
- Bromide (Br):≤500 ppm
Specification Test Results
Frequently Bought Together