Order Entry
Italy
ContactUsLinkComponent
4,4-Difluorocyclohexanone ≥97%
Irritant
4,4-Difluorocyclohexanone ≥97%
Catalog # H26027.03
CAS Number:  
Irritant
4,4-Difluorocyclohexanone ≥97%
Catalog # H26027.03
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

About this item

 :
 :134.12 g/mol
Melting Pt:34…40 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:22515-18-0
Formula:C₆H₈F₂O
MDL Number:MFCD08146635