Order Entry
Italy
ContactUsLinkComponent
4-(2,4-Dichlorophenoxy)butyric acid (2,4-DB)
4-(2,4-Dichlorophenoxy)butyric acid (2,4-DB)
Catalog # EHERC11980000
Supplier:  EHRENSTORFER
CAS Number:  
4-(2,4-Dichlorophenoxy)butyric acid (2,4-DB)
Catalog # EHERC11980000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
EINECS:202-366-9
CAS Number:94-82-6
Formula:C₁₀H₁₀Cl₂O₃
MDL Number:MFCD00002819