3-Methyl-1,2-cyclopentanedione for synthesis, Sigma-Aldrich®
8.14675.1000
: Merck
:
- Pack type:Plastic bottle for solids
- Pk:1 kg
- Seq:4024
- Solubility:(20 °C) soluble
- Melting Point:104 - 107 °C
- Molar Mass:112.13 g/mol
- Flash Point:100 °C
- Hill Formula:C6H8O2
:
:112.13 g/mol
:210 °C (1013 hPa)
:M-Clarity™ quality level = MQ200
:Refrigerator
:201-303-2
:765-70-8
:C₆H₈O₂
:MFCD00001417