Order Entry
Italy
Orders LinkContactUsLinkComponent
Woulff bottles
Woulff bottles
Supplier:   witeg Labortechnik
Woulff bottles
Supplier:  witeg Labortechnik
Certificates
Woulff bottles
4 Options Available Below
Product Details & Documents
Made of borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion gives relatively high resistance to temperature changes
With three necks.
Product Family Options
Product Information
  • Capacity
    Ext.Ø
    Height
    Neck
    Availability
    Price
  • 500 ml
    85 mm
    155 mm
    with NS joint
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 135,00 €
    135,00 €
    1 * 1 pezzi
       135,00 €
  • 2000 ml
    130 mm
    225 mm
    with NS joint
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 218,00 €
    218,00 €
    1 * 1 pezzi
       218,00 €
  • 5000 ml
    185 mm
    290 mm
    with NS joint
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 333,00 €
    333,00 €
    1 * 1 pezzi
       333,00 €
  • 10000 ml
    225 mm
    345 mm
    with NS joint
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 503,00 €
    503,00 €
    1 * 1 pezzi
       503,00 €