Screw caps, glass filled, WHEATON®
Catalog # WHEAW224600
Supplier: DWK Life Sciences
Specifications
- Cap Size:24-400
- Pk:200
Specifications
About this item
Open top polypropylene glass-filled cap.
- Provides greater heat resistance than other PP caps
- PTFE / silicone septa bonded to cap
- Ideal for EPA samples