Calcein
Supplier: Thermo Fisher Scientific
About this item
Synonyms: Oftasceina, Fluorescein Complexon, Oftasceinum, 2,7-Bis[bis(carboxymethyl)aminomethyl]fluorescein, Oftasceine, 2,2',2'',2'''-(((3',6'-Dihydroxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-2',7'-diyl)bis(methylene))bis(azanetriyl))tetraacetic acid, Fluorexon, Bis[N,N-bis(carboxymethyl)aminomethyl]fluorescein, NSC 298193
- CAS Number:1461-15-0
- Formula:C₃₀H₂₆N₂O₁₃
- MDL Number:MFCD00005049
- Molecular Weight:622.53 g/mol
- Melting Pt:200 °C
- Storage Temperature:Ambient
- EINECS:215-957-1
2 Options Available Below
Product Details & Documents
Synonyms: Oftasceina, Fluorescein Complexon, Oftasceinum, 2,7-Bis[bis(carboxymethyl)aminomethyl]fluorescein, Oftasceine, 2,2',2'',2'''-(((3',6'-Dihydroxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-2',7'-diyl)bis(methylene))bis(azanetriyl))tetraacetic acid, Fluorexon, Bis[N,N-bis(carboxymethyl)aminomethyl]fluorescein, NSC 298193
- CAS Number:1461-15-0
- Formula:C₃₀H₂₆N₂O₁₃
- MDL Number:MFCD00005049
- Molecular Weight:622.53 g/mol
- Melting Pt:200 °C
- Storage Temperature:Ambient
- EINECS:215-957-1
Specifications for Product Family
Specification Test Results
- Appearance:Yellow to orange-brown Powder
- Infrared spectrum:Conforms
- UV spectrum:Complies
- Suitability:(for Ca determination) Passes test
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



