Order Entry
Italy
ContactUsLinkComponent
Nalidixic acid
Nalidixic acid
Catalog # EHERC15412000
Supplier:  EHRENSTORFER
CAS Number:  
Nalidixic acid
Catalog # EHERC15412000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g
  • Seq:
    3020

Specifications

About this item

 :
 :232.24 g/mol
EINECS:206-864-7
 :GHS08
 :GHS07
Storage Temperature:Ambient
CAS Number:389-08-2
Formula:C₁₂H₁₂N₂O₃
MDL Number:MFCD00006884