Order Entry
Italy
ContactUsLinkComponent
 
 
3-(2-Benzimidazolyl)-7-(diethylamino)coumarin 98%, Sigma-Aldrich®
  416541-100MG
 :  
undefined
 
3-(2-Benzimidazolyl)-7-(diethylamino)coumarin 98%, Sigma-Aldrich®
  416541-100MG
 :  

 

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg
  • Seq:
    2440

 

 

  • Appearance (Colour):
    Yellow to Gold
  • Appearance (Form):
    Powder
  • Infrared spectrum:
    Conforms to Structure
  • Carbon:
    70.2 - 73.9 %
  • Nitrogen:
    12.3 - 12.9 %
  • Purity (HPLC):
    ≥97.5 %

 

 

 :
 :333.3838 g/mol
 :234...237 °C (lit.)
 :248-451-4
 :GHS07
 :Room Temperature
 :27425-55-4
 :C₂₀H₁₉N₃O₂
 :MFCD00051348