L(-)-Carnitine 99+%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: Levocarnitine, L-Carnitine, Vitamin BT, (R)-3-Hydroxy-4-(trimethylammonio)butanoate, R)-(-)-3-Hydroxy-4-(trimethylammonio)butyrate
- CAS Number:541-15-1
- Formula:C₇H₁₅NO₃
- MDL Number:MFCD00038747
- Molecular Weight:161.20 g/mol
- Melting Pt:208…212 °C (dec.)
- Storage Temperature:Refrigerator
- EINECS:208-768-0
3 Options Available Below
Product Details & Documents
Synonyms: Levocarnitine, L-Carnitine, Vitamin BT, (R)-3-Hydroxy-4-(trimethylammonio)butanoate, R)-(-)-3-Hydroxy-4-(trimethylammonio)butyrate
- CAS Number:541-15-1
- Formula:C₇H₁₅NO₃
- MDL Number:MFCD00038747
- Molecular Weight:161.20 g/mol
- Melting Pt:208…212 °C (dec.)
- Storage Temperature:Refrigerator
- EINECS:208-768-0
Specifications for Product Family
Specification Test Results
- Appearance:White Crystalline powder or crystals
- Infrared spectrum:Conforms
- Solubility:(5 % in water) Clear colorless
- Specific optical rotation:-29° to -32° (20°C, 589 nm) (c=1, H2O)
- Titration with HClO4:≥99.0 %
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



