Order Entry
Italy
ContactUsLinkComponent
Organic reference standard, Benzo[a]fluoranthene 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Organic reference standard, Benzo[a]fluoranthene 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL20560000CY
Supplier:  EHRENSTORFER
CAS Number:  
Organic reference standard, Benzo[a]fluoranthene 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL20560000CY
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    203-33-8
  • Description:
    Benzo[a]fluoranthene 10 µg/ml in cyclohexane
  • Formula:
    C₂₀H₁₂
  • Description:
    Benzo[a]fluoranthene

Specifications

About this item