Order Entry
Italy
ContactUsLinkComponent
Propanephosphonic acid cyclic anhydride 50% (w/w) in ethyl acetate
Irritant
Corrosive
Flammable
Propanephosphonic acid cyclic anhydride 50% (w/w) in ethyl acetate
Catalog # ACRO209010500
CAS Number:  
undefined
Irritant
Corrosive
Flammable
Propanephosphonic acid cyclic anhydride 50% (w/w) in ethyl acetate
Catalog # ACRO209010500
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    50 mL
  • Seq:
    2370

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to amber Solution
  • Assay:
    ≥50 % w/w Solids

Specification Test Results

About this item

 :
 :318.19 g/mol
Boiling Pt:77.2 °C
Flash Pt:–5 °C
Storage Temperature:Ambient
EINECS:422-210-5
UN:2924
ADR:3,II
 :GHS07
 :GHS05
 :GHS02
CAS Number:68957-94-8
Formula:C₉H₂₁O₆P₃
MDL Number:MFCD00006583