Order Entry
Italy
ContactUsLinkComponent
5-Bromo-4-chloro-3-indolyl phosphate disodium salt (BCIP-Na) ≥98%
5-Bromo-4-chloro-3-indolyl phosphate disodium salt (BCIP-Na) ≥98%
Catalog # H56265.03
CAS Number:  
5-Bromo-4-chloro-3-indolyl phosphate disodium salt (BCIP-Na) ≥98%
Catalog # H56265.03
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

About this item

CAS Number:102185-33-1
 :
Formula:C₈H₄BrClNNa₂O₄P
MDL Number:MFCD00036757
 :370.43 g/mol