2,4-Diquinolinecarboxylic acid for synthesis, Sigma-Aldrich®
Catalog # 8.15103.0025
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Melting Point:229 °C
- Molar Mass:217.18 g/mol
Specification Test Results
About this item
:
:M-Clarity™ quality level = MQ200
CAS Number:5323-57-9
Formula:C₁₁H₇NO₄