Order Entry
Italy
ContactUsLinkComponent
4,4'-Dimethoxytrityl chloride 98%
Irritant
4,4'-Dimethoxytrityl chloride 98%
Certificates
undefined
Irritant
4,4'-Dimethoxytrityl chloride 98%

Specification Test Results

  • Appearance:
    Pink Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    121 to 127 °C
  • Titration Argentometric:
    ≥97.5 % (ex Cl)

Specification Test Results

Related Information

Frequently Bought Together