Allyl acetoacetate for synthesis, Sigma-Aldrich®
8.18694.0100
: Merck
:
- Pack type:Glass bottle
- Pk:100 ml
- Seq:2440
- Solubility:48 g/l (20 °C)
- Molar Mass:142.15 g/mol
- Boiling Point:87 - 91 °C (13 hPa)
- Vapor Pressure:0.2 hPa (20 °C)
- Flash Point:67 °C
- Refractive Index:1.438 (25 °C, 589 nm)
- Explosion Limit:1.15 %(V)
- Density:1.038 g/cm3 (20 °C)
- pH:3.7 (48 g/l, H2O, 20 °C)
- Ignition Point:300 °C
:
:142.1525 g/mol
:6.1,III
:M-Clarity™ quality level = MQ200
:214-269-9
:2810
:GHS06
:Ambient
:1118-84-9
:C₇H₁₀O₃
:MFCD00009811