Alcian blue 8GX, certified
Supplier: Thermo Fisher Scientific
About this item
- CAS Number:33864-99-2
- Formula:C₅₆H₆₈Cl₄CuN₁₆S₄
- MDL Number:MFCD00010720
- Molecular Weight:1298.86 g/mol
- Melting Pt:148 °C
- Storage Temperature:Ambient
- EINECS:251-705-7
3 Options Available Below
Product Details & Documents
- CAS Number:33864-99-2
- Formula:C₅₆H₆₈Cl₄CuN₁₆S₄
- MDL Number:MFCD00010720
- Molecular Weight:1298.86 g/mol
- Melting Pt:148 °C
- Storage Temperature:Ambient
- EINECS:251-705-7
Documents
ACRO40046
SDS
Specifications for Product Family
Specification Test Results
- Appearance:Dark blue to purple Powder
- Infrared spectrum:Conforms
- Dye content:≥50 %
- UV Lambda max:615 to 635 nm (in water)
- BSC Certification:Passes
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



