Imidazole-4,5-dicarboxylic acid for synthesis, Sigma-Aldrich®
8.18370.0010
: Merck
:
- Pack type:Plastic bottle
- Pk:10 g
- Seq:3236
- Solubility:0.5 g/l (20 °C)
- Melting Point:270 - 275 °C
- Molar Mass:156.1 g/mol
- Bulk Density:650 - 660 kg/m3
- pH:3.7 (0.5 g/l, H2O, 20 °C)
- Ignition Point:430 °C
- Hill Formula:C5H4N2O4
:
:M-Clarity™ quality level = MQ200
:156.10 g/mol
:209-327-5
:570-22-9
:C₅H₄N₂O₄
:MFCD00005200