Order Entry
Italy
ContactUsLinkComponent
2-(Trimethylsiloxy)-1,3-butadiene for synthesis, Sigma-Aldrich®
Flammable
2-(Trimethylsiloxy)-1,3-butadiene for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Flammable
2-(Trimethylsiloxy)-1,3-butadiene for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Molar Mass:
    142.27 g/mol
  • Boiling Point:
    124 °C (1013 hPa)
  • Flash Point:
    12 °C
  • Density:
    0.82 g/cm3 (20 °C)
  • Ignition Point:
    350 °C

Specification Test Results

Related Information

Frequently Bought Together