5-Fluoro-2'-deoxyuridine (floxuridine), Millipore®
Catalog # 343333-50
Supplier: Merck Millipore (Calbiochem)
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:50 mg
- Seq:2370
Specifications
About this item
:
:246.19 g/mol
EINECS:200-072-5
UN:2811
ADR:6.1,III
CAS Number:50-91-9
Formula:C₉H₁₁FN₂O₅
MDL Number:MFCD00006530