Order Entry
Italy
ContactUsLinkComponent
(Bromomethyl)triphenylphosphonium bromide 98%
Irritant
(Bromomethyl)triphenylphosphonium bromide 98%
Certificates
undefined
Irritant
(Bromomethyl)triphenylphosphonium bromide 98%

Specification Test Results

  • Appearance:
    White to light beige to pink Powder or crystals
  • Infrared spectrum:
    Conforms
  • Melting point:
    232 to 242 °C
  • Titration Argentometric:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together