Order Entry
Italy
ContactUsLinkComponent
Solvent ASTM D2896, Chlorobenzene/Acetic acid-mixture (2/1 v/v), TBN solvent for synthesis, Sigma-Aldrich®
Flammable
Corrosive
Irritant
Environment
Solvent ASTM D2896, Chlorobenzene/Acetic acid-mixture (2/1 v/v), TBN solvent for synthesis, Sigma-Aldrich®
Catalog # 8.19145.2500
Supplier:  Merck
CAS Number:  
Flammable
Corrosive
Irritant
Environment
Solvent ASTM D2896, Chlorobenzene/Acetic acid-mixture (2/1 v/v), TBN solvent for synthesis, Sigma-Aldrich®
Catalog # 8.19145.2500
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

About this item

 :
Boiling Pt:∼118 °C (1013 hPa)
Density:∼1.08 g/cm³ (20 °C)
UN:3265
ADR:8,II
 :M-Clarity™ quality level = MQ200
Flash Pt:32 °C
 :GHS02
 :GHS05
 :GHS07
 :GHS09
Storage Temperature:Ambient