Sodium methoxide ∼25% (w/w) in methanol
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:124-41-4
:
Formula:CH₃NaO
MDL Number:MFCD00012179
:54.02 g/mol
Merck Index:14,8643
Boiling Pt:92 °C
Melting Pt:–2 °C
Density:0.945
Flash Pt:29 °C (84 °F)
EINECS:204-699-5
UN:1289
ADR:3,III
:GHS06
:GHS08
:GHS05
:GHS02
Storage Temperature:Ambient
Related Information
Frequently Bought Together