

Terephthalic acid ≥99.0% (by GC, titration analysis)
: TCI
:100-21-0
:
:C₈H₆O₄
:MFCD00002558
:166.13 g/mol
:14,09162
:392 °C (1013 hPa)
:1,472 g/cm³ (20 °C)
:260 °C
:202-830-0
:GHS07
:GHS08
:Ambient
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 99.0 %
- Purity (Neutralization titration):min. 99.0 %
- Solubility in 1mol/L NaOH:almost transparency
Frequently Bought Together