



Sodium bis(trimethylsilyl)amide 1 M in THF
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1070-89-9
:
Formula:C₆H₁₈NNaSi₂
MDL Number:MFCD00009835
:183.38 g/mol
Density:0.904
Flash Pt:–17 °C (1 °F)
Storage Temperature:Ambient
EINECS:213-983-8
UN:2924
ADR:3,II
:GHS02
:GHS05
:GHS08
:GHS07
Related Information
Frequently Bought Together