Order Entry
Italy
ContactUsLinkComponent
Sudan black B, certified
Sudan black B, certified
Catalog # ACRO419830100
CAS Number:  
undefined
Sudan black B, certified
Catalog # ACRO419830100
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 g
  • Seq:
    3236
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Dark brown to black Crystalline powder
  • Infrared spectrum:
    Conforms
  • UV Lambda max:
    596 to 605 nm
  • Absorption Ratio P-15/P+15:
    0.98 to 1.02
  • BSC Certification:
    Passes

Specification Test Results

About this item

Synonyms: CI 26150, Ceres black BN, Solvent Black 3, Fat Black HB
 :
 :456.55 g/mol
Melting Pt:120…124 °C
Storage Temperature:Ambient
EINECS:224-087-1
CAS Number:4197-25-5
Formula:C₂₉H₂₄N₆
MDL Number:MFCD00006919