Soybean trypsin inhibitor, MP Biomedicals
Trypsin inhibitor is a reversible serine protease inhibitor. It inhibits factor Xa, trypsin, chymotrypsin, kallikrein and plasmin.
Unit definition: One unit will inhibit one trypsin BAEE unit. One trypsin unit causes an increase of absorbance at 253 nm of 0,001 per minute at 25 °C.
- CAS No.:9035-81-8
- Species:Soy bean
- Storage conditions:−20 °C
- Protein/peptide name:Trypsin inhibitor
- Molecular weight:28,5 kDa
Frequently Bought Together