
(-)-Dibenzoyl-L-tartaric acid monohydrate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:62708-56-9
:
Formula:C₁₈H₁₄O₈·1H₂O
MDL Number:MFCD00149119
:376.33 g/mol (358.31 anhy)
Melting Pt:88…90 °C
EINECS:220-374-0
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Powder
- Infrared spectrum:Conforms
- Water:4 to 5.5 %
- HPLC:≥97.5 %
- Specific optical rotation:-113° to -117° (20°C, 589 nm) (c=5, ethanol)
- Titration with TBAH:97.5 to 102.5 %
Specification Test Results
Frequently Bought Together