Leupeptin hemisulphate, MP Biomedicals
Supplier: MP Biomedicals
Certificates
About this item
Leupeptin is a modified tripeptide reversible protease inhibitor of trypsin-like proteases and some cysteine proteases including endoproteinase Lys-C, kallikrein, papain, thrombin, cathepsin B, cathepsins H and L, trypsin, calpain, trypsin8 and plasmin.
- Soluble in water (50 mg/ml)
- White powder
- Effective concentration: 10 to 100 μM
It gives multiple peaks on HPLC due to equilibria among three forms in solution. Purity determined using three main peaks. Majority of contaminating peptide is racemised leupeptin.
Specifications
- CAS No.:103476-89-7
- Storage conditions:−20 °C
- MDL No.:MFCD00037012
- Protein/peptide name:Leupeptin hemisulphate
- Molecular weight:475,6 kDa
- Molecular formula:C₂₀H₃₈N₆O₄ · ½ H₂SO₄
Specifications
Frequently Bought Together