Order Entry
Italy
ContactUsLinkComponent
E-64 ((2S,3S)-(+)-trans-Epoxy succinyl-L-leucylamido-4-guanidino butane), MP Biomedicals
E-64 ((2S,3S)-(+)-trans-Epoxy succinyl-L-leucylamido-4-guanidino butane), MP Biomedicals
Supplier:  MP Biomedicals
Certificates
E-64 ((2S,3S)-(+)-trans-Epoxy succinyl-L-leucylamido-4-guanidino butane), MP Biomedicals
Supplier:  MP Biomedicals

Specifications

  • CAS No.:
    66701-25-5
  • Storage conditions:
    −20 °C
  • MDL No.:
    MFCD00080261
  • Protein/peptide name:
    E-64 ((2S,3S)-(+)-trans-Epoxy succinyl-L-leucylamido-4-guanidino butane)
  • Molecular weight:
    357,4 kDa
  • Molecular formula:
    C₁₅H₂₇N₅O₅

Specifications

Frequently Bought Together