Adenosine-5'-diphosphoric acid (ADP)
Catalog # ALPC101312
Supplier: Alpha Laboratories
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1,5 mL
- Seq:2095
Specifications
About this item
:
:427.2 g/mol
EINECS:200-392-5
CAS Number:58-64-0
Formula:C₁₀H₁₅N₅O₁₀P₂
MDL Number:MFCD00066473