2,4-Dichloro-6,7-dimethoxyquinazoline
Catalog # PROOMM0085.06
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:259.0887 g/mol
CAS Number:27631-29-4
Formula:C₁₀H₈Cl₂N₂O₂
MDL Number:MFCD00051733