
2,4-Dichloro-6,7-dimethoxyquinazoline 97%
:27631-29-4
:
:C₁₀H₈Cl₂N₂O₂
:MFCD00051733
:259.0887 g/mol
:172…177 °C
:GHS07
:Ambient
- Appearance:White to yellow to beige Crystalline powder
- Infrared spectrum:Conforms
- Melting point:172 to 179 °C
- HPLC:≥96.0 %
Frequently Bought Together